EDTA tetrasodium salt

Supplier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Ethylenediaminetetraacetic acid tetrasodium saltTetrasodium ethylenediaminetetraacetateEthylenedinitrilotetra-acetic acid tetrasodium saltEDTA tetrasodiumEdetic acid tetrasodium saltKestranal® 4S

ROTH3619.1EA 0 GBP
ROTH3619.1
EDTA tetrasodium salt
EDTA tetrasodium salt
Formula: C₁₀H₁₂N₂Na₄O₈
MW: 380.17 g/mol
Melting Pt: > 300 °C
Density: 6.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150027
CAS Number: 64-02-8
EINECS: 200-573-9
Merck Index: 12,03557
https://uk.vwr.com/stibo/web/std.lang.all/59/84/2355984.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR