N-Vanillylnonanamide [=Capsaicin (Synthetic)] ≥96% (by HPLC), white to off-white solid
Supplier: ENZO LIFE SCIENCES AG
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
N-Pelargonylvanillylamide
, N-Nonylvanylamide
, Nonivamide
, N-Pelargonic acid Vanillylamide
, N-(4-Hydroxy-3-methoxybenzyl)nonanamide
, Capsaicin (Synthetic)
ENZOALX550239M100EA
56.2
GBP
ENZOALX550239M100
N-Vanillylnonanamide [=Capsaicin (Synthetic)] ≥96% (by HPLC), white to off-white solid
N-Vanillylnonanamide [=Capsaicin (Synthetic)]
Minor constituent of hot pepper (capsicum oleoresin).
Soluble in DMSO or 100% ethanol; slightly soluble in water.
Formula:
C₁₇H₂₇NO₃ MW: 293.41 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00017286 CAS Number: 2444-46-4 |
Specification Test Results
Purity (HPLC) | ≥96% |
Identity | Determined by NMR |
Source | Synthetic |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...