Me-7BIO ≥98%

Supplier: ENZO LIFE SCIENCES AG
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: (2’Z,3’E)-7-Bromo-1-methylindirubin-3’-oxime

ENZOALX430157M001EA 154 GBP
ENZOALX430157M001
Me-7BIO ≥98%
Me-7BIO

Control compound for 6BIO.


Soluble in DMSO.

Order Now

Specification Test Results

Purity ≥98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR