Cobalt(III) protoporphyrin IX chloride ≥95% (by TLC)

Supplier: ENZO LIFE SCIENCES AG
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

ENZOALX430076M025EA 158 GBP
ENZOALX430076M025
Cobalt(III) protoporphyrin IX chloride ≥95% (by TLC)
Cobalt(III) protoporphyrin IX chloride

Heme oxygenase activator.


The ability of cobalt ions to induce heme oxygenase (HO) activity in liver cells was first reported almost 30 years ago. Cobalt protoporphyrin IX (CoPP) was later shown to be a substrate for HO, now identified as the inducible isoform HO-1 (heast shock protein 32; hsp32). CoPP has been shown to down-regulate various cytochrome P450 isoforms and various mechanisms of action have been attributed to CoPP and other metalloporphyrins.


Soluble in basic aqueous solutions at pH between 8.5 to 9.0. Once in solution pH can be adjusted to physiological pH.

Formula: C₃₄H₃₂CoN₄O₄·Cl
MW: 654.5 g/mol
Storage Temperature: Freezer
CAS Number: 102601-60-5

Order Now

Specification Test Results

Purity (TLC) ≥95%
Identity Determined by UV/VIS, NMR and MS

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR