Ingenol ≥98%, high purity

Supplier: ENZO LIFE SCIENCES AG
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (1aR,2S,5R,5aR,6S,8aS,9R,10aR)-5,5a,6-Trihydroxy-4-(hydroxymethyl)-1,1,7,9-tetramethyl-1a,2,5,5a,6,9,10,10a-octahydro-1H-2,8a-methanocyclopenta[a]cyclopropa[e][10]annulen-11-one

ENZOALX350031M005EA 331 GBP
ENZOALX350031M005
Ingenol ≥98%, high purity
Ingenol

PKC inhibitor


Extremely weak protein kinase C (PKC) activator. Starting material for synthesis of ingenol derivatives.

Formula: C₂₀H₂₈O₅
MW: 348.43 g/mol
MDL Number: MFCD27980538
CAS Number: 30220-46-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR