myo-Inositol
Supplier: ENZO LIFE SCIENCES AG
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Inositol
, rel-(1R,2r,3S,4R,5s,6S)-Cyclohexane-1,2,3,4,5,6-hexaol
, D-Myo-Inositol
ENZOALX307003G025EA
51.8
GBP
ENZOALX307003G025
myo-Inositol
myo-Inositol
Formula:
C₆H₁₂O₆ MW: 180.16 g/mol Boiling Pt: 291 °C (1013 hPa) Melting Pt: 210 °C Density: 1,752 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00077932 CAS Number: 87-89-8 EINECS: 201-781-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...