(R)-Ibuprofen ≥98%

Supplier: ENZO LIFE SCIENCES AG
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (R)-2-(4-Isobutylphenyl)propanoic acid(R)-(-)-Ibuprofen

ENZOBMLEI2900200EA 157 GBP
ENZOBMLEI2900200
(R)-Ibuprofen ≥98%
(R)-Ibuprofen

Less active enantiomer of ibuprofen.


  • COX inhibitor


White crystalline powder.

Formula: C₁₃H₁₈O₂
MW: 206.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00069290
CAS Number: 51146-57-7
https://uk.vwr.com/stibo/web/std.lang.all/37/41/45403741.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR