Geldanamycin ≥98% Biotin
Supplier: ENZO LIFE SCIENCES AG
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ENZOBMLEI3410001EA
279
GBP
ENZOBMLEI3410001
Geldanamycin ≥98% Biotin
Geldanamycin
A useful tool for affinity purification of HSP-90 and HSP-90 dependent client proteins.
- HSP90 inhibitor
Formula:
C₂₉H₄₀N₂O₉ MW: 560.64 g/mol |
MDL Number:
MFCD00274570 CAS Number: 30562-34-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...