Anti-MLKL Mouse Monoclonal Antibody (DyLight 594) [clone: 954724]

Supplier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

NOVUMAB91871DL594EA 0 GBP
NOVUMAB91871DL594
Anti-MLKL Mouse Monoclonal Antibody (DyLight 594) [clone: 954724]
Antibodies
Type:
Antigen: MLKL
Clonality: Monoclonal
Clone: 954724
Conjugation: DyLight 594
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR