Kanamycin sulphate

Supplier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Kanamycin MonosulphateKanamycin A

02EFC06191EA 0 GBP
02EFC06191
Kanamycin sulphate
Kanamycin sulphate

Kanamycin sulfate is a broad spectrum aminoglycoside-antibiotic derived from Streptomyces kanamyceticus. It is used as an additive in culture media for the isolation of group D streptococci on Kanamycin Esculin Azide Agar and for selection of transformed plant cells containing the neomycin phosphotransferase on a kanamycin-medium. Kanamycin sulfate can also be used as a selection agent for cells transformed with kanamycin B resistance gene.


  • Antibiotic
  • Inhibitor of protein synthesis


Caution: Research or further manufacturing use only, not for direct human use.

Formula: C₁₈H₃₈N₄O₁₅S
MW: 582.6 g/mol
Melting Pt: >250 °C
MDL Number: MFCD00070253
CAS Number: 25389-94-0

Order Now

Specification Test Results

Identity Test Passes
Potecy 782 µg/mg (as is)
790 µg/mg (dry)
Crystallinity Passes
pH (1% aq soln) 8.2
Loss on Drying 1.03%
Residue on Ignition 0.22%
Chromatographic Purity Passes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR