Hydrochloric acid 0.01 mol/l (0.01 N) Reag. Ph. Eur. 1043504

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Muriatic acidHydrochloric acid solution

85849.290EA 59.3 GBP
85849.290
Hydrochloric acid 0.01 mol/l (0.01 N) Reag. Ph. Eur. 1043504
Hydrochloric acid

Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formula: HCl
MW: 36.46 g/mol
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,III

For metal trace analysis in AAS or ICP

Ask for your “customized multi-element standard solution” with your exact composition using the request form

Ask for a quotation

12/31/2022

Order Now

For metal trace analysis in AAS or ICP

Ask for your “customized multi-element standard solution” with your exact composition using the request form

Ask for a quotation

12/31/2022

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR