Dichloromethane for HPLC

Supplier: MARKET SOURCE PART PROCESS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Methylene chloride

XXXXRH1010.2500EA 32.6 GBP
XXXXRH1010.2500 MSPPRH1010.0500S XXXXRH1010
Dichloromethane for HPLC
Dichloromethane
Formula: CH₂Cl₂
MW: 84.93 g/mol
Boiling Pt: 39.8 °C (1013 hPa)
Melting Pt: –95 °C
Density: 1.322 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1,III
Merck Index: 12,06140
https://uk.vwr.com/stibo/web/std.lang.all/14/30/20961430.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR