Chloramphenicol, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Chloromycetin®(R,R)-ChloramphenicolTega-Cetin

M-Clarity™ quality level = MQ100

31667-250MGEA 37.5 GBP
31667-250MG
Chloramphenicol, Supelco®
Chloramphenicol
Formula: Cl₂CHCONHCH(CH₂OH)CH(OH)C₆H₄NO₂
MW: 323.13 g/mol
Boiling Pt: 645 °C (1013 hPa)
Melting Pt: 150 °C
Density: 1.49 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00078159
CAS Number: 56-75-7
EINECS: 200-287-4
Merck Index: 13,02087

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR