4-Cholesten-3-one
Supplier: EHRENSTORFER GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(+)-4-Cholesten-3-one
, (8S,9S,10R,13R,14S,17R)-10,13-Dimethyl-17-((R)-6-methylheptan-2-yl)-6,7,8,9,10,11,12,13,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3(2H)-one
EHERC11665300EA
206
GBP
EHERC11665300
4-Cholesten-3-one
4-Cholesten-3-one
Formula:
C₂₇H₄₄O MW: 384.65 g/mol Melting Pt: 81…82 °C |
MDL Number:
MFCD00003663 CAS Number: 601-57-0 EINECS: 210-005-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...