4-Cholesten-3-one

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (+)-4-Cholesten-3-one(8S,9S,10R,13R,14S,17R)-10,13-Dimethyl-17-((R)-6-methylheptan-2-yl)-6,7,8,9,10,11,12,13,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3(2H)-one

MOLE18488191-25GEA 0 GBP
MOLE18488191-25G
4-Cholesten-3-one
4-Cholesten-3-one
Formula: C₂₇H₄₄O
MW: 384.65 g/mol
Melting Pt: 81…82 °C
MDL Number: MFCD00003663
CAS Number: 601-57-0
EINECS: 210-005-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR