3-Pyridineboronic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Pyridinylboronic acid, Pyridin-3-ylboronic acid, Pyridine-3-boronic acid

010819-5G 010819-25G
FLUO010819-5GEA 0 GBP
FLUO010819-5G FLUO010819-25G
3-Pyridineboronic acid
3-Pyridineboronic acid
Formula: C₅H₆BNO₂
MW: 122.92 g/mol
MDL Number: MFCD00674177
CAS Number: 1692-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR