5-Bromo-2,3-diaminopyridine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Bromopyridine-2,3-diyldiamine, 2,3-Diamino-5-bromopyridine

M-Clarity™ quality level = MQ200

551899-25G 551899-5G
SIAL551899-25GEA 150 GBP
SIAL551899-25G SIAL551899-5G
5-Bromo-2,3-diaminopyridine, Sigma-Aldrich®
5-Bromo-2,3-diaminopyridine
Formula: C₅H₆BrN₃
MW: 188.03 g/mol
Boiling Pt: 321 °C (1013 hPa)
MDL Number: MFCD00460094
CAS Number: 38875-53-5
EINECS: 254-172-9
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR