Cholesterol 95% stabilised

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: CholesterinCholest-5-en-3-ol(3Beta)-Cholest-5-en-3-ol

ACRO110195000EA 193 GBP
ACRO110195000 ACRO110190250 ACRO110191000 ACRO110190025
Cholesterol 95% stabilised
Cholesterol
Formula: C₂₇H₄₆O
MW: 386.66 g/mol
Boiling Pt: 360 °C (1013 hPa)
Melting Pt: 147…150 °C
Density: 1.06 g/cm³
Flash Pt: 250 °C
Storage Temperature: Ambient
MDL Number: MFCD00003646
CAS Number: 57-88-5
EINECS: 200-353-2
Merck Index: 13,02221

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
GC ≥94 %
Stabilizer 50 to 350 ppm alpha-Tocopherol
GC ≥98.5 %
Acid value ≤1 mg KOH/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR