2-Fluoro-6-phenoxybenzonitrile
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
006319-5G
006319-1G
006319-25G
FLUO006319-5GEA
0
GBP
FLUO006319-5G
FLUO006319-1G
FLUO006319-25G
2-Fluoro-6-phenoxybenzonitrile
2-Fluoro-6-phenoxybenzonitrile
Formula:
C₁₃H₈FNO Boiling Pt: 338 °C (1013 hPa) Melting Pt: 90…93 °C |
MDL Number:
MFCD00068201 CAS Number: 175204-06-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...