2-Fluoro-6-methoxybenzonitrile
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CFF623.5
006266-25G
006266-100G
CHMPCFF623.5EA
0
GBP
CHMPCFF623.5
FLUO006266-25G
FLUO006266-100G
2-Fluoro-6-methoxybenzonitrile
2-Fluoro-6-methoxybenzonitrile
Formula:
C₈H₆FNO MW: 151.14 g/mol Boiling Pt: 245 °C (1013 hPa) Melting Pt: 93…95 °C |
MDL Number:
MFCD00042291 CAS Number: 94088-46-7 EINECS: 302-047-5 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...