2-Amino-3,5-dibromobenzonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CFF403.10 CFF403.25 CFF403.50 017426-10G 017426-25G 017426-50G 017426-5G
CHMPCFF403.10EA 0 GBP
CHMPCFF403.10 CHMPCFF403.25 CHMPCFF403.50 FLUO017426-10G FLUO017426-25G FLUO017426-50G FLUO017426-5G
2-Amino-3,5-dibromobenzonitrile
2-Amino-3,5-dibromobenzonitrile
Formula: C₇H₄Br₂N₂
MW: 275.93 g/mol
MDL Number: MFCD00179571
CAS Number: 68385-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR