Arsenic(III) oxide ≥99.0%, ReagentPlus®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: di-Arsenic trioxide

SIALA1010-25GEA 0 GBP
SIALA1010-25G
Arsenic(III) oxide ≥99.0%, ReagentPlus®
Arsenic (III) oxide
Formula: As₂O₃
MW: 197.84 g/mol
Boiling Pt: 465 °C (1013 hPa)
Melting Pt: 312 °C
Density: 3.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003433
CAS Number: 1327-53-3
EINECS: 215-481-4
UN: 1561
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR