Resistivity Kit amd Boost for High-Flow Systems
Supplier: Millipore
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MILLZKITRES01SDEA 1030 GBP
MILLZKITRES01SD
Resistivity Kit amd Boost for High-Flow Systems
Resistance Testers
This Resistivity boost kit includes both resistivity monitor and a booster. The resistivity monitor installed within the water system verifies resistivity levels of water in the distribution loop.
  • Ergonomic and patented pack-locking mechanism
The resistivity booster ensures the correct resistivity water flows in the loop and into the storage tanks. It may be easily installed on the front of the water purification system. The ergonomic and patented pack-locking mechanism allows for rapid replacement when needed. This kit is designed for use with Milli-Q® HX 7000 SD water systems.