Human HSP90 ELISA Kit (Chemiluminescence)

Supplier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

NOVUNBP2-76643EA 0 GBP
NOVUNBP2-76643
Human HSP90 ELISA Kit (Chemiluminescence)
Assays ELISAs
This kit recognizes Human HSP-90 in samples. No significant cross-reactivity or interference between Human HSP-90 and analogues was observed.

  • Standard curve range: 62,5 to 4000 pg/ml
  • Inter-Assay: CV% <10,47%
  • Intra-Assay: CV% <9,68%
  • Spike Recovery: 91 to 117%
  • Gene: HSP90AA1

Hsp90 has at least 2 isoforms, Hsp90a and Hsp90b which are encoded by separate genes. These ubiquitous and highly conserved proteins account for 1 to 2% of all cellular proteins in most cells. Hsp90 is part of the cells powerful network of chaperones to fight the deleterious consequences of protein unfolding caused by nonphysiological conditions. However, in the absence of stress, Hsp90 is a necessary component of fundamental cellular processes such as hormone signaling and cell cycle control. In this context several key regulatory proteins such as steroid receptors, cell cycle kinases involved in signal transduction and p53 have been identified as substrates of Hsp90. It has been suggested that Hsp90 acts as a capacitor for morphological evolution by buffering widespread variation, which may affect morphogenic pathways.

Delivery information: Kit contains Biotinylated detection Ab diluent, Concentrated biotinylated detection Ab (100x), Concentrated HRP conjugate (100x), Concentrated wash buffer (25x), HRP conjugate diluent, Micro CLIA plate (dismountable), Plate sealer, Product manual, Reference standard, Sample diluent, Substrate reagent A and B.

Caution: For research use only and is not approved for use in humans or in clinical diagnosis.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR