Ultra-Micro SpinColumns
Supplier: Harvard Apparatus
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HARV747212EA 1720 GBP
HARV747212
Ultra-Micro SpinColumns
Filters Centrifugal Filters
Ultra-Micro SpinColumns are best suited for sample volumes from 10 to 25 µl. Centrifugation or filtration under vacuum or pressure can be used to run the sample through the columns.
  • Rapid sample preparation time
  • Micro sample sizes
  • High sample recovery
Results obtained from the Micro SpinColumns are highly reproducible and there is minimal sample loss. Each column includes two 2 ml centrifuge tubes plus top caps (for Gel Filtration) or frit.
Applications: DNA, protein and peptide purification, small molecule, carbohydrate, salt and radiolabel removal, Nick translation, affinity separation, buffer exchange.