trans-Anethole, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(E)-1-Methoxy-4-(prop-1-en-1-yl)benzene
M-Clarity™ quality level = MQ200
117870-25ML
117870-100ML
SIAL117870-25MLEA
62.9
GBP
SIAL117870-25ML
SIAL117870-100ML
trans-Anethole, Sigma-Aldrich®
trans-Anethole
Formula:
C₁₀H₁₂O MW: 148.2 g/mol Boiling Pt: 236 °C (1013 hPa) Melting Pt: 21…23 °C Density: 0.9883 g/cm³ (20 °C) Flash Pt: 96 °C |
MDL Number:
MFCD00009284 CAS Number: 4180-23-8 EINECS: 224-052-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...