Boron trichloride 1 mol/l in hexane, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

211249-100ML 211249-800ML
SIAL211249-100MLEA 113 GBP
SIAL211249-100ML SIAL211249-800ML
Boron trichloride 1 mol/l in hexane, Sigma-Aldrich®
Boron trichloride
Formula: BCl₃
MW: 117.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011313
CAS Number: 10294-34-5
EINECS: 233-658-4
UN: 3286
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR