4-Bromothiazole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

016178-5G 016178-1G
FLUO016178-5GEA 0 GBP
FLUO016178-5G FLUO016178-1G
4-Bromothiazole
4-Bromothiazole
Formula: C₃H₂BrNS
MW: 164.03 g/mol
MDL Number: MFCD06657592
CAS Number: 34259-99-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR