3-Acetylphenylboronic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3'-Boronoacetophenone, 3-Acetylphenylboronic Acid, 3-Acetylbenzeneboronic acid, Dihydroxy(3-acetylphenyl)borane, (3-Acetylphenyl)metaboric acid, 3-Acetylphenylboronic acid, 3-Acetylbenzeneboronic acid

021605-10G 021605-25G 021605-5G
FLUO021605-10GEA 0 GBP
FLUO021605-10G FLUO021605-25G FLUO021605-5G
3-Acetylphenylboronic acid
3-Acetylphenylboronic acid
Formula: C₈H₉BO₃
MW: 163.97 g/mol
MDL Number: MFCD01074678
CAS Number: 204841-19-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR