(±)-β-Citronellol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: β-Citronellol3,7-Dimethyloct-6-en-1-olCitronellol(±)-CitronellolDL-Citronellol

M-Clarity™ quality level = MQ300

SIALW230901-4KG-KEA 229 GBP
SIALW230901-4KG-K
(±)-β-Citronellol, Sigma-Aldrich®
(±)-β-Citronellol
Formula: C₁₀H₂₀O
MW: 156.27 g/mol
Boiling Pt: 222 °C (1013 hPa)
Density: 0.86 g/cm³ (20 °C)
Flash Pt: 100 °C
MDL Number: MFCD00002935
CAS Number: 106-22-9
EINECS: 203-375-0
UN: 3082
ADR: 9,III
Merck Index: 13,02354

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR