Anethole

Supplier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: cis-Anethol, 1-Methoxy-4-(prop-1-en-1-yl)benzene

5131.1 5131.2
ROTH5131.1EA 20.58 GBP
ROTH5131.1 ROTH5131.2
Anethole
Anethole
Formula: C₁₀H₁₂O
MW: 148.2 g/mol
Boiling Pt: 234.5 °C (1013 hPa)
Melting Pt: 21.3 °C
Density: 1 g/cm³ (12 °C)
MDL Number: MFCD00009284
CAS Number: 104-46-1
EINECS: 203-205-5
Merck Index: 13,00647

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR