4-Chloro-3,5-xylenol, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Chloroxylenol
, 4-Chloro-3,5-dimethylphenol
, 2-Chloro-5-hydroxy-3-xylene
M-Clarity™ quality level = MQ200
8.18734.0250EA
49.9
GBP
8.18734.0250
8.18734.9999
4-Chloro-3,5-xylenol, Sigma-Aldrich®
4-Chloro-3,5-xylenol
Formula:
C₈H₉ClO MW: 156.61 g/mol Boiling Pt: 246 °C (1013 hPa) Melting Pt: 114…116 °C Density: 1.364 g/cm³ (20 °C) Flash Pt: 132 °C |
MDL Number:
MFCD00002324 CAS Number: 88-04-0 EINECS: 201-793-8 Merck Index: 13,02194 |
Specification Test Results
Solubility | 250 mg/l (20 °C) |
Melting Point | 114 - 116 °C |
Molar Mass | 156.61 g/mol |
Boiling Point | 246 °C (1013 hPa) |
Flash Point | 132 °C |
Density | 0.67 g/cm3 |
pH | 7 (0.2 g/l, H2O) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...