4,5-Dimethoxy-2-nitrobenzyl alcohol

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 6-Nitroveratryl alcohol, (4,5-Dimethoxy-2-nitrophenyl)methanol

D4595.5 D4595.25
XXXXD4595.5EA 0 GBP
XXXXD4595.5 XXXXD4595.25
4,5-Dimethoxy-2-nitrobenzyl alcohol
4,5-Dimethoxy-2-nitrobenzyl alcohol
Formula: C₉H₁₁NO₅
MW: 213.19 g/mol
MDL Number: MFCD00014701
CAS Number: 1016-58-6
EINECS: 213-806-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR