N,N'-Dimethylethylendiamine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: N1,N2-Dimethylethane-1,2-diamine, sym. Dimethylethylendiamine, N,N'-Dimethylethylenediamine

M-Clarity™ quality level = MQ100

270032-100G 270032-25G
SIAL270032-100GEA 189 GBP
SIAL270032-100G SIAL270032-25G
N,N'-Dimethylethylendiamine, Sigma-Aldrich®
N,N'-Dimethylethylendiamine
Formula: C₄H₁₂N₂
MW: 88.15 g/mol
Boiling Pt: 119 °C (1013 hPa)
Density: 0.82 g/cm³ (20 °C)
MDL Number: MFCD00008290
CAS Number: 110-70-3
EINECS: 203-793-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR