(4-Fluorophenyl)acetyl chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Fluorobenzeneacetyl chloride, 4-Fluorophenylacetyl Chloride

009229-1G 009229-5G
FLUO009229-1GEA 0 GBP
FLUO009229-1G FLUO009229-5G
(4-Fluorophenyl)acetyl chloride
(4-Fluorophenyl)acetyl chloride
Formula: (C₆H₄F)CH₂COCl
MW: 172.59 g/mol
Boiling Pt: 202…204 °C (1013 hPa)
Density: 1.259 g/cm³ (20 °C)
MDL Number: MFCD00674181
CAS Number: 459-04-1
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR