N,N'-Ethylenedianiline, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
N1,N2-Diphenylethane-1,2-diamine, 1,2-Dianilinoethane, N,N'-Diphenylethylenediamine
M-Clarity™ quality level = MQ100
D27004-25G
D27004-5G
SIALD27004-25GEA
110
GBP
SIALD27004-25G
SIALD27004-5G
N,N'-Ethylenedianiline, Sigma-Aldrich®
N,N'-Ethylenedianiline
Formula:
C₆H₅NHCH₂CH₂NHC₆H₅ MW: 212.29 g/mol Boiling Pt: 228 °C (1013 hPa) Melting Pt: 68 °C |
MDL Number:
MFCD00003019 CAS Number: 150-61-8 EINECS: 205-765-6 Merck Index: 13,03011 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...