1-Pentyl-4-iodobenzene

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Iodopentylbenzene, 4-Pentyliodbenzol, 1-(4-Iodophenyl)pentane, 1-Iodo-4-pentylbenzene, 4-Pentyliodobenzene, 1-Iodo-4-n-pentylbenzene

I1211.5 I1211.25
XXXXI1211.5EA 0 GBP
XXXXI1211.5 XXXXI1211.25
1-Pentyl-4-iodobenzene
1-Pentyl-4-iodobenzene
Formula: C₁₁H₁₅I
MW: 274.14 g/mol
MDL Number: MFCD00079767
CAS Number: 85017-60-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR