1-(4-Fluorophenyl)pyrrole

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: N-(4-Fluorophenyl)pyrrole, 1-(4-Fluorophenyl)-1H-pyrrole

F1531-G.1 F1531-G.5
XXXXF1531-G.1EA 0 GBP
XXXXF1531-G.1 XXXXF1531-G.5
1-(4-Fluorophenyl)pyrrole
1-(4-Fluorophenyl)pyrrole
Formula: C₁₀H₈FN
MW: 161.18 g/mol
MDL Number: MFCD00047066
CAS Number: 81329-31-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR