(3,5-Dimethoxyphenyl)acetonitrile

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3,5-Dimethoxyphenylacetonitrile, 3,5-Dimethoxybenzyl cyanide, 2-(3,5-Dimethoxyphenyl)acetonitrile

D4741.5 D4741.25
XXXXD4741.5EA 0 GBP
XXXXD4741.5 XXXXD4741.25
(3,5-Dimethoxyphenyl)acetonitrile
(3,5-Dimethoxyphenyl)acetonitrile
Formula: C₁₀H₁₁NO₂
MW: 177.2 g/mol
Boiling Pt: 298 °C (1013 hPa)
Melting Pt: 53 °C
MDL Number: MFCD00016395
CAS Number: 13388-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR