3-(Diethylamino)propyl isothiocyanate

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Diethylamino-1-propyl isothiocyanate, 3-(Diethylamino)propylisothiocyanate

D2745.5 D2745.25
XXXXD2745.5EA 0 GBP
XXXXD2745.5 XXXXD2745.25
3-(Diethylamino)propyl isothiocyanate
3-(Diethylamino)propyl isothiocyanate
Formula: C₈H₁₆N₂S
MW: 172.29 g/mol
Boiling Pt: 88…92 °C (0,1 mmHg)
Density: 0,9535 g/cm³ (20 °C)
MDL Number: MFCD00041133
CAS Number: 2626-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR