5-Chloro-2-thenoic acid

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Chlorothiophene-2-carboxylic acid, 5-Chloro-2-thiophenecarboxylic acid

C3042.5 C3042.25
XXXXC3042.5EA 0 GBP
XXXXC3042.5 XXXXC3042.25
5-Chloro-2-thenoic acid
5-Chloro-2-thenoic acid
Formula: C₅H₃ClO₂S
MW: 162.6 g/mol
MDL Number: MFCD00041426
CAS Number: 24065-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR