4-Chlorobenzyl isothiocyanate

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Chloro-4-(isothiocyanatomethyl)benzene

C1840.5 C1840.25
XXXXC1840.5EA 0 GBP
XXXXC1840.5 XXXXC1840.25
4-Chlorobenzyl isothiocyanate
4-Chlorobenzyl isothiocyanate
Formula: C₈H₆ClNS
MW: 183.66 g/mol
Boiling Pt: 105…108 °C (0,3 mmHg)
Density: 1,291 g/cm³ (20 °C)
MDL Number: MFCD00041116
CAS Number: 3694-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR