3-Chlorobenzyl isothiocyanate

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

C1839.5 C1839.25
XXXXC1839.5EA 0 GBP
XXXXC1839.5 XXXXC1839.25
3-Chlorobenzyl isothiocyanate
3-Chlorobenzyl isothiocyanate
Formula: C₈H₆ClNS
MW: 183.66 g/mol
MDL Number: MFCD00046820
CAS Number: 3694-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR