4-Isothiocyanatobenzoic acid

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

C1324.5 C1324.25
XXXXC1324.5EA 0 GBP
XXXXC1324.5 XXXXC1324.25
4-Isothiocyanatobenzoic acid
4-Isothiocyanatobenzoic acid
Formula: C₈H₅NO₂S
MW: 179.2 g/mol
MDL Number: MFCD00041369
CAS Number: 2131-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR