3-Isothiocyanatobenzoic acid

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Carboxyphenyl isothiocyanate

C1323.5 C1323.25
XXXXC1323.5EA 0 GBP
XXXXC1323.5 XXXXC1323.25
3-Isothiocyanatobenzoic acid
3-Isothiocyanatobenzoic acid
Formula: C₈H₅NO₂S
MW: 179.2 g/mol
Boiling Pt: 336 °C (1013 hPa)
Melting Pt: 164…165 °C
MDL Number: MFCD00041367
CAS Number: 2131-63-7
UN: 2923
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR