5-Bromo-2-thenoic acid

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Bromothiophene-2-carboxylic acid, 5-Bromo-2-thiophenecarboxylic acid

B3293.5 B3293.25
XXXXB3293.5EA 0 GBP
XXXXB3293.5 XXXXB3293.25
5-Bromo-2-thenoic acid
5-Bromo-2-thenoic acid
Formula: C₅H₃BrO₂S
MW: 207.05 g/mol
MDL Number: MFCD00079725
CAS Number: 7311-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR