1-Adamantyl isothiocyanate

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

A1543.2 A1543.10
XXXXA1543.2EA 0 GBP
XXXXA1543.2 XXXXA1543.10
1-Adamantyl isothiocyanate
1-Adamantyl isothiocyanate
Formula: C₁₁H₁₅NS
MW: 193.31 g/mol
Boiling Pt: 270 °C (1013 hPa)
Melting Pt: 174…175 °C
MDL Number: MFCD00074736
CAS Number: 4411-26-1
EINECS: 224-564-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR