2-Acetyl-5-bromothiophene

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(5-Bromo-2-thienyl)-1-ethanone

A1380.10 A1380.50
XXXXA1380.10EA 0 GBP
XXXXA1380.10 XXXXA1380.50
2-Acetyl-5-bromothiophene
2-Acetyl-5-bromothiophene
Formula: C₆H₅BrOS
MW: 205.08 g/mol
Boiling Pt: 253 °C (1013 hPa)
Melting Pt: 92…93 °C
MDL Number: MFCD00014528
CAS Number: 5370-25-2
EINECS: 226-363-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR