Benzyl propionate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ300

W215015-100G-K W215015-1KG-K
SIALW215015-100G-KEA 193 GBP
SIALW215015-100G-K SIALW215015-1KG-K
Benzyl propionate, Sigma-Aldrich®
Benzyl propionate
Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 222 °C (1013 hPa)
Density: 1.029…1.033 g/cm³ (20 °C)
MDL Number: MFCD00027003
CAS Number: 122-63-4
EINECS: 204-559-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR