2-Thiophenemethanol, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Thiophen-2-methanol, Thiophen-2-ylmethanol, 2-Thienylmethanol
M-Clarity™ quality level = MQ100
T32654-100G
T32654-25G
SIALT32654-100GEA
153
GBP
SIALT32654-100G
SIALT32654-25G
2-Thiophenemethanol, Sigma-Aldrich®
2-Thiophenemethanol
Formula:
C₅H₆OS MW: 114.17 g/mol Boiling Pt: 207 °C (1013 hPa) Melting Pt: 0 °C Density: 1.206 g/cm³ (20 °C) |
MDL Number:
MFCD00005454 CAS Number: 636-72-6 EINECS: 211-264-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...